Gel and PCR Clean-up, NucleoSpin®
Supplier: MACHEREY-NAGEL
Certificates
About this item
NucleoSpin® Gel and PCR Clean-up kits allow 2-in-1 clean up with optimised recovery and elution volume.
- Two applications in one kit – one buffer with optimal performance for both PCR clean-up and gel extraction
- High recovery with low elution volumes – efficient purification of fragments down to 50 bp
- Minimised elution volume of 15 μl – highly concentrated DNA
- Binding buffer with pH indicator – easy visualisation of correct binding conditions
Applications include purification of PCR products, extraction of DNA from agarose gels, DNA extraction from polyacrylamide gels. Typical downstream applications include cloning, sequencing, PCR, restriction analysis.
NucleoSpin® Gel and PCR Clean-up ensures complete removal of contaminations such as nucleotides, primers; enzymes; mineral oil; PCR additives (eg. salts, betaine, DMSO); detergents (eg. Tween 20, Triton X-100); dyes (eg. ethidium bromide, crystal violet, Stain G, Midori Green, Roti®-Safe GelStain, DNA SafeStain).
Delivery: Each kit contains NucleoSpin® Gel and PCR Clean-up columns, collection tubes (2 ml) and buffers.
Frequently Bought Together