GC capillary column connectors, Swagelok®
Supplier: Restek
Certificates
About this item
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation or replacing worn connections.
- Fittings are available in brass or stainless steel
- Treated with Siltek®/Sulfinert or Silcosteel®-CR
Frequently Bought Together