Boc-L-Arg(Z)2-OH ≥98.0% (by HPLC), Novabiochem®
Catalog # 04-12-0220-5
Supplier: Novabiochem (Merck Biosciences)
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:542.6 g/mol
Storage Temperature:Freezer
CAS Number:51219-19-3
Formula:C₂₇H₃₄N₄O₈
MDL Number:MFCD00236824