Fittings, secure-fit
Supplier: Restek
Certificates
About this item
Secure-Fit provides a good connection between HPLC components for trouble-free chromatography.
- Consistent, leak-free seal
- Eliminates the excess dead volume
Internal spring mechanism holds the capillary tubing at the proper depth in the female fitting. This seal is maintained while the nut is finger tightened.
Frequently Bought Together