ContactUsLinkComponent
Autosampler plunger carrier belt
Catalog # REST22695
Supplier:  Restek
undefined
Autosampler plunger carrier belt
Catalog # REST22695
Supplier:  Restek
Specifications
  • Description:
    Autosampler plunger carrier belt
  • Pk:
    1
Specifications
About this item
Autosampler plunger carrier belt for Agilent 7673A and 7673B autosamplers.