ContactUsLinkComponent
Injector wrench for Agilent
Injector wrench for Agilent
Catalog # REST22065
Supplier:  Restek
undefined
Injector wrench for Agilent
Catalog # REST22065
Supplier:  Restek
Specifications
  • Description:
    Injector wrench for Agilent 5890/6890/6850 GCs
  • Material:
    Stainless steel
  • Pk:
    1
Specifications
About this item
This stainless steel injector wrench is uses to remove the septum nut and weldments during GC maintenance.
  • Smaller end to remove the septum nut
  • Larger end to tighten the split/splitless weldment nut