Order Entry
Austria
ContactUsLinkComponent
Brain-derived Neurotrophic Factor (BDNF) Rapid™ ELISA Kit (CE marked)
Brain-derived Neurotrophic Factor (BDNF) Rapid™ ELISA Kit (CE marked)
Supplier:  Biosensis
Certificates
undefined
Brain-derived Neurotrophic Factor (BDNF) Rapid™ ELISA Kit (CE marked)
Supplier:  Biosensis

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Target protein:
    BDNF
  • Cross reactivity:
    Reacts with BDNF from mouse, rat and numerous other species due to BDNF sequence homology
  • Detection range:
    7,8 - 500 pg/ml
  • Storage temperature:
    2...8 °C
  • Sensitivity:
    Typical limit of detection (LOD) for BDNF is < 3 pg/mL determined by calculating the mean plus 2× standard deviation of mean of blank (n=20)

Specifications

Frequently Bought Together