Orders LinkContactUsLinkComponent
Drum racks
Drum racks
Supplier:   CEMO
Drum racks
Supplier:  CEMO
Certificates
Drum racks
16 Options Available Below
Product Details & Documents
Drum racks for approved and space saving storage of barrels and other containers.
  • All drum racks are completely hotdip galvanised and have a corrosion free GRP sump pallet for long-term safe use
  • With general construction inspection approval no. Z-40.12-227 granted by DIBt-Berlin
  • Universally applicable for waste oil, lyes and acids
Product Family Options
Product Information
  • Weight
    W×D×H
    Availability
    Price
  • CEMG7081
    Type 120, with GRP sump pallet (65 l), drum support for 2×60 l barrels, heights of storage levels from ground 630/1150 mm
    Type 120, with GRP sump pallet (65 l), drum support for 2×60 l barrels, heights of storage levels from ground 630/1150 mm
    18 kg
    450×820×1310 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 493,00 €
    493,00 €
    1 * 1 ST
       493,00 €
  • CEMG7640
    Type 120, with GRP sump pallet (65 l), drum and grating support for 1×60 l barrel and small containers, heights of storage levels from ground 600/1120 mm
    Type 120, with GRP sump pallet (65 l), drum and grating support for 1×60 l barrel and small containers, heights of storage levels from ground 600/1120 mm
    19 kg
    450×820×1310 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 537,00 €
    537,00 €
    1 * 1 ST
       537,00 €
  • CEMG7639
    Type 120, with GRP sump pallet (65 l) and grating support for small containers, heights of storage levels from ground 600/1120 mm
    Type 120, with GRP sump pallet (65 l) and grating support for small containers, heights of storage levels from ground 600/1120 mm
    21 kg
    450×820×1310 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 552,00 €
    552,00 €
    1 * 1 ST
       552,00 €
  • CEMG7642
    Type 360 with GRP sump pallet (150 l), drum support for 6×60 l barrels and small containers, heights of storage levels from ground 690/1170 mm
    Type 360 with GRP sump pallet (150 l), drum support for 6×60 l barrels and small containers, heights of storage levels from ground 690/1170 mm
    60 kg
    1320×800×1300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 738,00 €
    738,00 €
    1 * 1 ST
       738,00 €
  • CEMG10253
    Type 360, with steel sump pallet (SW2), drum support for 6×60 l barrels and small containers, hights of storage levels from ground 360/690/1170 mm
    Type 360, with steel sump pallet (SW2), drum support for 6×60 l barrels and small containers, hights of storage levels from ground 360/690/1170 mm
    115 kg
    1320×800×1300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 878,00 €
    878,00 €
    1 * 1 ST
       878,00 €
  • CEMG7663
    Type 400, with GRP sump pallet (220 l), barrel support for 2×60 l or 1×200 l barrels and small containers, heights of storage levels from ground 620 mm
    Type 400, with GRP sump pallet (220 l), barrel support for 2×60 l or 1×200 l barrels and small containers, heights of storage levels from ground 620 mm
    41 kg
    990×1280×840 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 673,00 €
    673,00 €
    1 * 1 ST
       673,00 €
  • CEMG7665
    Type 400, with GRP sump pallet (220 l), barrel support for 2×60 l or 1×200 l barrels and grating support for small containers, heights of storage levels from ground 620/1390 mm
    Type 400, with GRP sump pallet (220 l), barrel support for 2×60 l or 1×200 l barrels and grating support for small containers, heights of storage levels from ground 620/1390 mm
    82 kg
    990×1280×1620 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 020,00 €
    1 020,00 €
    1 * 1 ST
       1 020,00 €
  • CEMG7664
    Type 400, with GRP sump pallet (220 l), barrel support for 4×60 l or 2×200 l barrels or 2×60 l and 1×200 l drums for small containers, heights of storage levels from ground 620/1400 mm
    Type 400, with GRP sump pallet (220 l), barrel support for 4×60 l or 2×200 l barrels or 2×60 l and 1×200 l drums for small containers, heights of storage levels from ground 620/1400 mm
    70 kg
    990×1280×1620 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 050,00 €
    1 050,00 €
    1 * 1 ST
       1 050,00 €
  • CEMG7734
    Type 540, with GRP sump pallet (150 l), drum support for 6×60 l barrels and small containers, heights of storage levels from ground 690/1170/1650 mm
    Type 540, with GRP sump pallet (150 l), drum support for 6×60 l barrels and small containers, heights of storage levels from ground 690/1170/1650 mm
    82 kg
    1320×800×2000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 907,00 €
    907,00 €
    1 * 1 ST
       907,00 €
  • CEMG10254
    Type 540, with steel sump pallet (SW2), drum support for 6×60 l barrels and small containers, heights of storage levels from ground 360/690/1170/1650 mm
    Type 540, with steel sump pallet (SW2), drum support for 6×60 l barrels and small containers, heights of storage levels from ground 360/690/1170/1650 mm
    137 kg
    1320×800×2000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 090,00 €
    1 090,00 €
    1 * 1 ST
       1 090,00 €
  • CEMG7673
    Type 800, with GRP sump pallet (220 l), barrel support for 2×200 l barrels and grating support for small containers, heights of storage levels from ground 620/1390 mm
    Type 800, with GRP sump pallet (220 l), barrel support for 2×200 l barrels and grating support for small containers, heights of storage levels from ground 620/1390 mm
    116 kg
    1410×1280×1620 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 250,00 €
    1 250,00 €
    1 * 1 ST
       1 250,00 €
  • CEMG7672
    Type 800, with GRP sump pallet (220 l), barrel support for 2×200 l and 3×60 l barrels and small containers, heights of storage levels from ground 620/1400 mm
    Type 800, with GRP sump pallet (220 l), barrel support for 2×200 l and 3×60 l barrels and small containers, heights of storage levels from ground 620/1400 mm
    87 kg
    1410×1280×1620 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 310,00 €
    1 310,00 €
    1 * 1 ST
       1 310,00 €
  • CEMG7670
    Type 800, with GRP sump pallet (220 l), barrel support for 2×200 l barrels and small containers, heights of storage levels from ground 620 mm
    Type 800, with GRP sump pallet (220 l), barrel support for 2×200 l barrels and small containers, heights of storage levels from ground 620 mm
    51 kg
    1410×1280×840 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 804,00 €
    804,00 €
    1 * 1 ST
       804,00 €
  • CEMG7671
    Drum rack, type 800, with GRP sump pallet (220 l), barrel support for 4×200 l barrels and small containers, heights of storage levels from ground 620/1400 mm
    Drum rack, type 800, with GRP sump pallet (220 l), barrel support for 4×200 l barrels and small containers, heights of storage levels from ground 620/1400 mm
    84 kg
    1410×1280×1620 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 310,00 €
    1 310,00 €
    1 * 1 ST
       1 310,00 €
  • CEMG7674
    Type 800, with GRP sump pallet (220 l), drum support for 3×60 l barrels and small containers, 620 mm, heights of storage levels from ground 620 mm
    Type 800, with GRP sump pallet (220 l), drum support for 3×60 l barrels and small containers, 620 mm, heights of storage levels from ground 620 mm
    54 kg
    1410×1280×840 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 834,00 €
    834,00 €
    1 * 1 ST
       834,00 €
  • CEMG7675
    Type 800, with GRP sump pallet (220 l), drum support for 3×60 l barrels and small containers, heights of storage levels from ground 620/1400 mm
    Type 800, with GRP sump pallet (220 l), drum support for 3×60 l barrels and small containers, heights of storage levels from ground 620/1400 mm
    90 kg
    1410×1280×1620 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 290,00 €
    1 290,00 €
    1 * 1 ST
       1 290,00 €
Accessories
Product Information
  • Catalog #
    Weight
    Availability
    Price
  • Can holder, galvanised, for drum racks type 120/360/540, with surface 230×110 mm
    Can holder, galvanised, for drum racks type 120/360/540, with surface 230×110 mm
    CEMG5917
    2 kg
    Log in for availability
    1 * 1 ST - 41,00 €
Product Information
  • Catalog #
    Weight
    Availability
    Price
  • Rotating roller support for drum racks type 400/800 (for 200 l barrel (pair), heights of storage levels 660 mm from ground
    Rotating roller support for drum racks type 400/800 (for 200 l barrel (pair), heights of storage levels 660 mm from ground
    CEMG7669
    6 kg
    Log in for availability
    1 * 1 ST - 235,00 €
Product Information
  • Catalog #
    Weight
    Availability
    Price
  • Drum supports for drum racks for type 360/540, 3 pieces for one storage level for optimum emptying of the 60 l drums
    Drum supports for drum racks for type 360/540, 3 pieces for one storage level for optimum emptying of the 60 l drums
    CEMG8254
    1 kg
    Log in for availability
    1 * 3 ST - 51,30 €
Product Information
  • Catalog #
    Weight
    Availability
    Price
  • Steel grating for drum racks, as 3rd (for type 360) or 4th storage level (for type 540), 1300×620×30 mm, 210 mm storage height from ground
    Steel grating for drum racks, as 3rd (for type 360) or 4th storage level (for type 540), 1300×620×30 mm, 210 mm storage height from ground
    CEMG6817
    18 kg
    Log in for availability
    1 * 1 ST - 127,00 €
Product Information
  • Catalog #
    Weight
    Availability
    Price
  • Can holder, galvanised, for drum racks type 400/800, with bearing surface 185×110 mm
    Can holder, galvanised, for drum racks type 400/800, with bearing surface 185×110 mm
    CEMG7727
    2 kg
    Log in for availability
    1 * 1 ST - 46,80 €
Product Information
  • Catalog #
    Weight
    Availability
    Price
  • Drum supports, 3 pieces per storage level, for drum racks type 360/540
    Drum supports, 3 pieces per storage level, for drum racks type 360/540
    CEMG7800
    1 kg
    Log in for availability
    1 * 1 ST - 48,30 €
Product Information
  • Catalog #
    Weight
    W×D×H
    Availability
    Price
  • Sump pallet, PE, 40 l, without grating
    Sump pallet, PE, 40 l, without grating
    CEMG7871
    5 kg
    1300×620×80 mm
    Log in for availability
    1 * 1 ST - 163,00 €