ContactUsLinkComponent
Microwave ovens, Phoenix
Microwave ovens, Phoenix
Catalog # CEMAS-924560
Supplier:  CEM
Microwave ovens, Phoenix
Catalog # CEMAS-924560
Supplier:  CEM
Specifications
  • Description:
    Microwave ashing system, Phoenix, 1400 W
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This rapid microwave ashing system consists of a microwave heating system containing a 5 L chamber which can be heated to 1000 °C or a 1,8 L chamber which can be heated to 1200 °C using microwave energy.
  • Internal temperature calibration software
  • Integral dual thermocouple Temperature Control System for either single set-point temperature or eight segmented ramping temperature control programs
  • Two RS232C 9-pin serial interfaces for option balance and external computer and one 25-pin parallel port for optional printer
The unit maintains 2,8 m³ of air flow per minute through the microwave cavity to rapidly ash materials and remove combustion by-products. Program storing, processing and reporting weight data and % Ash, % LOI, % ROI, % LOD, %Fixed, %Volatile and %Total Residue to the instrument display, printer or external computer.