Mobile sumps for storage of hazardous materials, GRP
Supplier: CEMO
Certificates
About this item
The mobile sumps are applicable for the storage and transport of environmentally hazardous substances.
- Sumps contains hot-dip galvanised grating and chassis with collision protection
- It has two steering and two fixed castors and a pushing bar for trouble-free steering
- With general construction inspection approval number Z-40.12-227
The thermosetting plastic base structure in conjugation with the glass reinforcement provide an advantageous fire behaviour and high chemical reistance.
Frequently Bought Together