Cold traps, with olives
Supplier: NEUBERT VOLUME GLASSWAERE
Certificates
About this item
This cooling traps are made from borosilicate glass 3.3, with sleeve on the lower part, two 10 mm glass poles and ground clamp made of PP.
Frequently Bought Together