Order Entry
Austria
ContactUsLinkComponent
Labelling tapes, Scotch® crystal
Labelling tapes, Scotch® crystal
Catalog # MMMAC6001933
Supplier:  3M
undefined
Labelling tapes, Scotch® crystal
Catalog # MMMAC6001933
Supplier:  3M

Specifications

  • Width:
    19,0 mm
  • Description:
    Labelling tapes, synthetic acrylate adhesive
  • Pk:
    24 Roll

Specifications

About this item

Scotch® crystal adhesive tape is a highly transparent and universally applicable adhesive tape of high quality. And the adhesive tape sticks reliably for sealing gifts and envelopes or for attaching light objects.

  • Adhesive tape fits into all refillable Scotch® dispensers
  • They are universal and indispensable tool for gluing
  • Tear and aging resistant

Length: 33,0 m.

Colour: Transparent.