Order Entry
Austria
ContactUsLinkComponent
Buffer, PBS (10X, Dulbecco's) powder pack
Buffer, PBS (10X, Dulbecco's) powder pack
Catalog # A0965.9010
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Buffer, PBS (10X, Dulbecco's) powder pack
Catalog # A0965.9010
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    PBS buffer (10X, Dulbecco's), 10 l

Specifications

About this item

Powder mixture to prepare PBS buffer (10X) according to Dulbecco.

Composed of di-sodium hydrogen phosphate anhydrous (11500 mg/L), potassium chloride (2000 mg/L), potassium dihydrogen phosphate (2000 mg/L) and sodium chloride (80000 mg/L).