Buffer solutions
Supplier: SI Analytics
Jump to:
About this item
Buffer solutions intended for extremely precise calibration.
- Highest measurement reliability
- No preservative agents
- Hermetically sealed in the glass ampoule
- Sterilised with hot steam
Certifications: According to DIN 19 266.
13 Options Available Below
Product Details & Documents
Buffer solutions intended for extremely precise calibration.
- Highest measurement reliability
- No preservative agents
- Hermetically sealed in the glass ampoule
- Sterilised with hot steam
Certifications: According to DIN 19 266.
Dokumente
Product Family Options
Product Information
- pH valuepH accuracyReference temperaturePkAvailabilityPrice
- SCOI285137977Type L4791
- SCOI285138213Type L4694
- SCOI285138632Type L4895/SET
- 662-6180Type L4698
- 662-0901Type L4794
- 662-1608Type L4790
- 662-1429Type L4798
- 662-6182Type L6874
- 662-0902Type L4796
- 662-6175Type L7004
- SCOI285138221Type L4697
- 662-0903Type L4799
- SCOI285138205Type L4691
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



