Orders LinkContactUsLinkComponent
Pycnometer with thermometer, adjusted
Pycnometer with thermometer, adjusted
Certificates
Pycnometer with thermometer, adjusted
Pycnometer with thermometer, adjusted
7 Options Available Below
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in capillary stopper
  • Very good chemical resistance
  • Very good temperature resistance
It includes pycnometer bottle and ground-in thermometer joint size 10/19 with blue filling.
Certifications: DIN ISO 3507
Product Family Options
Product Information
  • Volume (nom.)
    NS
    Availability
    Price
  • 25 ml
    10/19
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
  • 50 ml
    10/19
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €
  • 100 ml
    10/19
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 122,00 €
    122,00 €
    1 * 1 ST
       122,00 €
  • 10 ml
    10/19
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 151,00 €
    151,00 €
    1 * 1 ST
       151,00 €
  • 25 ml
    10/19
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 157,00 €
    157,00 €
    1 * 1 ST
       157,00 €
  • 50 ml
    10/19
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 164,00 €
    164,00 €
    1 * 1 ST
       164,00 €
  • 100 ml
    10/19
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 184,00 €
    184,00 €
    1 * 1 ST
       184,00 €