Order Entry
Austria
ContactUsLinkComponent
 
Hydrocarbon S-trap
Catalog # REST22016
Supplier:  Restek
undefined
Hydrocarbon S-trap
Catalog # REST22016
Supplier:  Restek

Specifications

  • Description:
    Hydrocarbon S-trap, 1/8" brass fittings
  • Pk:
    1

Specifications

About this item

This trap removes hydrocarbons and other contaminants. It fits in GC oven for easy thermal recharging.

  • Each individually activated to ensure maximum efficiency
  • Reduces organics to 0,1 ppm (assuming 100 ppm input)

Maximum operating pressure: 4,13 bar

Dimensions: 171×143 mm