Order Entry
Austria
ContactUsLinkComponent
Broom head, without handle
Broom head, without handle
Catalog # KING061024/00
Supplier:  KING DISPOSABLES
undefined
Broom head, without handle
Catalog # KING061024/00
Supplier:  KING DISPOSABLES

Specifications

  • Bristle colour:
    Natural
  • Description:
    Broom head
  • Head width:
    600 mm
  • Pk:
    1

Specifications

About this item

Suitable for large surfaces.

Length: 60 cm