ContactUsLinkComponent
D-Desthiobiotin
D-Desthiobiotin
Catalog # 71610-3
CAS Number:  
D-Desthiobiotin
Catalog # 71610-3
CAS Number:  
Specifications
  • Pack type:
    Plastic ampoule
  • Pk:
    1 g
  • Description:
    D-Desthiobiotin
Specifications
About this item
D-Desthiobiotin provides a gentle elution of StrepTag® II proteins from the biotin-binding site of StrepTactin® resins. D-Desthiobiotin is offered as a lyophilised powder for preparation of elution buffer.