Order Entry
Austria
ContactUsLinkComponent
 
Acetyl coenzyme A trilithium salt trihydrate, MP Biomedicals
 
Acetyl coenzyme A trilithium salt trihydrate, MP Biomedicals

 

  • CAS No.:
    75520-41-1
  • Storage conditions:
    –20 °C
  • Form:
    White powder
  • MDL No.:
    MFCD00167406
  • Enzyme name:
    Acetyl coenzyme A trilithium salt trihydrate
  • Enzyme synonyms:
    Acetyl-coenzyme-A-trilithium-S-trihydrate
  • Purity:
    92%
  • Molecular formula:
    C₂₃H₃₅Li₃N₇O₁₇P₃S·3H₂O

 

Frequently Bought Together