Order Entry
Austria
ContactUsLinkComponent
Anti-CD90.2 Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 30-H12]
Anti-CD90.2 Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 30-H12]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD90.2 Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 30-H12]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 65-0903-U100
  • 65-0903-U025
  • Antigen symbol:
  • Thy1.2
  • Thy1.2
  • Antigen name:
  • CD90.2
  • CD90.2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • Clone:
  • 30-H12
  • 30-H12
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 21838
  • 21838
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • 2-8°C protected from light

Specifications

Related Information

Frequently Bought Together