Anti-CD16 / CD32 Rat Monoclonal Antibody (Biotin) [clone: 2.4G2]
Supplier: Tonbo Biosciences
Certificates
About this item
The 2.4G2 antibody is specific for a common epitope found in the extracellular regions of mouse Fc-receptors Fc-gamma II (CD32) and Fc-gamma III (CD16). These receptors are expressed by B cells, monocytes, macrophages, NK cells, dendritic cells, and neutrophils.
Type: Primary
Antigen: CD16 / CD32
Clonality: Monoclonal
Clone: 2.4G2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Specifications
- Catalog No:
- 30-0161-U500
- 30-0161-U025
- 30-0161-U100
- Antigen symbol:
- FCGR3, IGFR3, FCGR2, IGFR2, FC Receptor
- FCGR3, IGFR3, FCGR2, IGFR2, FC Receptor
- FCGR3, IGFR3, FCGR2, IGFR2, FC Receptor
- Antigen name:
- CD16 / CD32
- CD16 / CD32
- CD16 / CD32
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Biotin
- Clone:
- 2.4G2
- 2.4G2
- 2.4G2
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Gene ID:
- 14130/14131
- 14130/14131
- 14130/14131
- Isotype:
- IgG2b
- IgG2b
- IgG2b
- Flow cytometry:
- Yes
- Yes
- Yes
- Storage temperature:
- 2-8°C protected from light
- 2-8°C protected from light
- 2-8°C protected from light
Specifications
Frequently Bought Together