Order Entry
Austria
ContactUsLinkComponent
Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Catalog # ICNA0210330401
Supplier:  MP Biomedicals
CAS Number:  
Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Catalog # ICNA0210330401
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    0,45 kg
  • Description:
    Yeast Hydrolysate, enzymatic, from Brewers Yeast
  • Seq:
    01

Specifications

About this item

Yeast Hydrolysate Enzymatic is an autolysed yeast extract produced from 100% brewers yeast. It is a concentrated source of soluble protein components, carbohydrates, and B complex vitamins natural to yeast.

  • Soluble in water
  • Standard plate count: <50,000/g
  • Protein (N×6,25): 50,0 to 65,0%