Orders LinkContactUsLinkComponent
Pneumatic lockouts, Quick-Disconnect
Pneumatic lockouts, Quick-Disconnect
Anbieter:   Brady
Pneumatic lockouts, Quick-Disconnect
Anbieter:  Brady
Certificates
Pneumatic lockouts, Quick-Disconnect
Pneumatic lockouts, Quick-Disconnect
2 Options Available Below
Product Details & Documents
These pneumatic quick-disconnect lockouts isolate pneumatic energy without the expense and inconvenience of installing in-line lockout valves.
  • Device is fitted to the male fitting, isolating equipment from all compressed air sources
  • Accommodates the majority of 6 mm, 9 mm and 12 mm male fittings
Max. shackle diameter: 7,84 mm
Min. shackle clearance: 20 mm
Product Family Options
Product Information
  • Colour
    Verfügbarkeit
    Preis
  • Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,10 €
    19,10 €
    1 * 1 ST
       19,10 €
  • Red
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 95,60 €
    95,60 €
    1 * 6 ST
       95,60 €