Rectangular semi-micro cuvettes
: Hellma
Rectangular semi-micro cuvettes with four polished windows and PTFE lids for fluorescence applications.
Rectangular semi-micro cuvettes with four polished windows and PTFE lids for fluorescence applications.
Product Family Options
- TypeHeightVolumePath lengthWindow material