Micro-volume syringes, positive displacement, (Hamilton)
Supplier: Restek
Certificates
About this item
These micro-volume syringes have samples contained in the needle with full plunger displacement, ensuring no dead volume.
- High precision and accuracy down to 0,1 µl
- Seal between plunger and needle can be tightened for high-pressure injection
Ordering information: Replaceable needle/plunger kits and barrels are available on request.
Frequently Bought Together