Orders LinkContactUsLinkComponent
Membrane potential detection kit, MITO-ID®
Membrane potential detection kit, MITO-ID®
Supplier:   Enzo Life Sciences
Membrane potential detection kit, MITO-ID®
Supplier:  Enzo Life Sciences
Certificates
Membrane potential detection kit, MITO-ID®
Membrane potential detection kit, MITO-ID®
2 Options Available Below
Product Details & Documents
MITO-ID® membrane potential detection kit monitors energetic status using a simple mix-and-read, no-wash protocol. It measures mitochondrial membrane potential with a cationic dye that fluoresces either green or orange depending upon membrane potential status.
  • Highly sensitive detection of mitochondrial membrane potential decline
  • Aids in the drug research process in preclinical drug safety assessment (ADME-Tox)
  • Suitable for chemical/environmental toxicity screening and characterisation
  • True mix-and-read homogeneous assay for live cells
  • Optimized for the measurement of mitochondria membrane potential (MMP) and cell viability by fluorescence microscopy and flow cytometry
Cell-based assays for evaluating the functional status of mitochondria are useful tools for elucidating the role of mitochondrial activity in drug-induced toxicity, the apoptosis cascade and other cellular and biochemical processes.
Delivery information: Kit contains MITO-ID® MP detection reagent, necrosis detection reagent, CCCP control, 10X assay buffer 1 and 50X assay buffer 2.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • ENZOENZ510180025
    MITO-ID® Membrane potential detection kit
    MITO-ID® Membrane potential detection kit
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 244,00 €
    244,00 €
    1 * 25 Tests
       244,00 €
  • ENZOENZ51018K100
    MITO-ID® membrane potential detection kit
    MITO-ID® membrane potential detection kit
    100 Tests
    Log in for availability
    Log in for availability
    1 * 100 Tests
    1 * 100 Tests - 674,00 €
    674,00 €
    1 * 100 Tests
       674,00 €