Orders LinkContactUsLinkComponent
Precision saccharometers
Supplier:   Ludwig Schneider
Precision saccharometers
Supplier:  Ludwig Schneider
Certificates
Precision saccharometers
7 Options Available Below
Product Details & Documents
Saccharometers for beer and wort according to Plato with blue filling thermometer.
  • Suitable for calibration
Error limit: ±0,1 °Plato
Reference temperature: 20 °C
 : Available with works calibration certificate or DAkkS calibration certificate on request.
 : In a square plastic sleeve.
Specifications for Product Family
Specifications
  • Catalog No:
  • SCNL3292109S
  • SCNL3292107S
  • SCNL3292110S
  • SCNL3292111S
  • SCNL3292108S
  • SCNL3292112S
  • SCNL3292113S
  • Reference temperature:
  • 20 °C
  • 20 °C
  • 20 °C
  • 20 °C
  • 20 °C
  • 20 °C
  • 20 °C
Specifications
Product Family Options
Product Information
  • Range
    Division
    Temperature Range
    Length
    Availability
    Price
  • 0 - 5 °P
    0,1°
    0…30 °C
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,20 €
    76,20 €
    1 * 1 ST
       76,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3292109S
    Reference temperature
    20 °C
  • 0 - 10 °P
    0,1°
    0…30 °C
    430 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3292107S
    Reference temperature
    20 °C
  • 5 - 10 °P
    0,1°
    0…30 °C
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,20 €
    76,20 €
    1 * 1 ST
       76,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3292110S
    Reference temperature
    20 °C
  • 10 - 15 °P
    0,1°
    0…30 °C
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,20 €
    76,20 €
    1 * 1 ST
       76,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3292111S
    Reference temperature
    20 °C
  • 10 - 20 °P
    0,1°
    0…30 °C
    430 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3292108S
    Reference temperature
    20 °C
  • 15 - 20 °P
    0,1°
    0…30 °C
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,20 €
    76,20 €
    1 * 1 ST
       76,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3292112S
    Reference temperature
    20 °C
  • 20 - 25 °P
    0,1°
    0…30 °C
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,20 €
    76,20 €
    1 * 1 ST
       76,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3292113S
    Reference temperature
    20 °C