ContactUsLinkComponent
Vu-tight installation fittings kit
Vu-tight installation fittings kit
Catalog # REST20504
Supplier:  Restek
undefined
Vu-tight installation fittings kit
Catalog # REST20504
Supplier:  Restek
Specifications
  • Description:
    Vu-tight installation fittings kit
  • Pk:
    1
Specifications
About this item
This kit includes a ¹/₄" stainless steel nut and graphite ferrule for attaching the liner to the GC inlet and a ¹/₄ to ¹/₁₆" stainless steel reducer, plus a ¹/₄" and 0,5 mm ID graphite ferrule for attaching the column to the liner.