Order Entry
Austria
ContactUsLinkComponent
Mouse cathepsin D ELISA kit
Mouse cathepsin D ELISA kit
Catalog # BIRBORB138062-96
Supplier:  Biorbyt
CAS Number:  
undefined
Mouse cathepsin D ELISA kit
Catalog # BIRBORB138062-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse Cathepsin D ELISA kit, 96-well
  • Detection range:
    156 - 10000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Mouse cathepsin D ELISA kit is used for quantitative detection of mouse cathepsin D in cell culture supernates, serum and plasma (heparin, EDTA).

Cathepsin D is a protein that in humans is encoded by the CTSD gene. This proteinase is a member of the peptidase C1 family, having a specificity similar to but narrower than that of pepsin A. It is mapped to 11p15.5.

The cDNA encodes a 412-amino acid protein with 20 and 44 amino acids in a pre- and prosegment, respectively. Cathepsin D is one of the lysosomal proteinases. It is expressed and is involved in proteolytic degradation, cell invasion, and apoptosis.