Order Entry
Austria
ContactUsLinkComponent
Human ZAG EIA kit
Human ZAG EIA kit
Catalog # BIRBORB78637-96
Supplier:  Biorbyt
CAS Number:  
Human ZAG EIA kit
Catalog # BIRBORB78637-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human ZAG EIA Kit
  • Detection range:
    0,1 - 1000 ng/ml
  • Sensitivity:
    21 pg/ml

Specifications

About this item

Human ZAG EIA kit is an in vitro quantitative assay for detecting zinc binding alpha 2 glycoprotein (ZAG) peptide using serum, plasma, culture supernatant and cell lysates.

Delivery: Kit includes ZAG microplate (96 wells) coated with secondary antibody, wash buffer concentrate 20X (25 ml), standard ZAG peptide (2×10 µl), anti-zag polyclonal antibody (2×5 µl), assay diluent A (30 ml), assay diluent B 5X (15 ml), biotinylated ZAG peptide (2×20 µl)), HRP-streptavidin concentrate 100X (600 µl), positive control (100 µl)), TMB one-step substrate reagent (12 ml), stop solution (8 ml), assay diagram, and user manual.