Order Entry
Austria
ContactUsLinkComponent
Human RBP-4 EIA kit
Human RBP-4 EIA kit
Catalog # BIRBORB78629-96
Supplier:  Biorbyt
CAS Number:  
Human RBP-4 EIA kit
Catalog # BIRBORB78629-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human RBP-4 EIA Kit
  • Detection range:
    0,1 - 1000 ng/ml
  • Sensitivity:
    460 pg/ml

Specifications

About this item

Human RBP-4 EIA kit is an in vitro quantitative assay for detecting human retinol binding protein 4 in serum, plasma, culture supernatant and cell lysates.

Delivery: Kit includes RBP4 microplate (96 well) coated with secondary antibody, wash buffer concentrate 20X (25 ml), standard RBP4 peptide (2×10 µl), assay diluent A (30 ml), assay diluent B 5X (15 ml), biotinylated peptide (2×20 µl), HRP-streptavidin 400X (600 µl), TMB one-step substrate (12 ml), stop solution (8 ml), assay diagram, positive control sample, capture antibody, user manual.