Orders LinkContactUsLinkComponent
Density hydrometers
Anbieter:   Ludwig Schneider
Density hydrometers
Anbieter:  Ludwig Schneider
Certificates
Density hydrometers
Density hydrometers
14 Options Available Below
Product Details & Documents
Density hydrometers calibrated at 20 °C.
* Works certificate.
 : Available with works or DAkkS calibration certificate on request.
 : Packed in a square plastic sleeve.
Product Family Options
Product Information
  • Range
    Division
    Length
    Verfügbarkeit
    Preis
  • 0,600 - 0,700 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,80 €
    15,80 €
    1 * 1 ST
       15,80 €
  • 0,700 - 0,800 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,70 €
    14,70 €
    1 * 1 ST
       14,70 €
  • 0,800 - 0,900 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,70 €
    14,70 €
    1 * 1 ST
       14,70 €
  • 0,900 - 1,000 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,70 €
    14,70 €
    1 * 1 ST
       14,70 €
  • 1,000 - 1,100 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,80 €
    15,80 €
    1 * 1 ST
       15,80 €
  • 1,100 - 1,200 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,80 €
    15,80 €
    1 * 1 ST
       15,80 €
  • 1,200 - 1,300 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,80 €
    15,80 €
    1 * 1 ST
       15,80 €
  • 1,300 - 1,400 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,10 €
    17,10 €
    1 * 1 ST
       17,10 €
  • 1,400 - 1,500 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,10 €
    17,10 €
    1 * 1 ST
       17,10 €
  • 1,500 - 1,600 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,10 €
    17,10 €
    1 * 1 ST
       17,10 €
  • 1,600 - 1,700 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,40 €
    18,40 €
    1 * 1 ST
       18,40 €
  • 1,700 - 1,800 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,40 €
    18,40 €
    1 * 1 ST
       18,40 €
  • 1,800 - 1,900 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,50 €
    20,50 €
    1 * 1 ST
       20,50 €
  • 1,900 - 2,000 g/ml
    0,001 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,50 €
    20,50 €
    1 * 1 ST
       20,50 €