Order Entry
Austria
ContactUsLinkComponent
Mouse major urinary protein 1 ELISA kit
Mouse major urinary protein 1 ELISA kit
Catalog # BIRBORB54814-96
Supplier:  Biorbyt
CAS Number:  
Mouse major urinary protein 1 ELISA kit
Catalog # BIRBORB54814-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse Major Urinary Protein 1 ELISA Kit
  • Detection range:
    0,625 - 40 ng/ml

Specifications

About this item

Mouse major urinary protein 1 ELISA kit for the quantification of mouse Mup1 in serum and plasma samples.

Major urinary protein 1 (Mup1), also known as Mup7, Up-1, Ltn-1, Mup-1, Mup-a, Mup10 and Lvtn-1, is a low molecular weight secreted protein produced predominantly from the liver. Structurally it belongs to the lipocalin family, which carries small hydrophobic ligands such as pheromones. It has been demonstrated that Mup1 is an important player in regulating energy expenditure and metabolism in mice. In both dietary and genetic obese mice, the circulating concentrations of Mup1 were markedly decreased compared with the lean controls. Replenishment of recombinant Mup-1 led to improved glucose tolerance and insulin sensitivity, as well as increased energy expenditure and locomotor activity in db/db diabetic mice. It was suggested that Mup1 also regulates systemic glucose and/or lipid metabolism through the paracrine/autocrine regulation of the hepatic gluconeogenic and/or lipogenic programs, respectively.

Delivery: Micro-titre strips (96 wells) coated with a polyclonal antibody against mouse Mup1, sealed, wash buffer (10×40 ml), assay buffer (5×20 ml), detection antibody solution, a polyclonal antibody against mouse Mup1 conjugated with horseradish peroxidise (100×0,12 ml), recombinant mouse Mup1 in a buffered protein base lyophilised (40 ng), substrate solution (12 ml), stop solution (12 ml).