Orders LinkContactUsLinkComponent
Nitrogen generators, precision nitrogen trace
Nitrogen generators, precision nitrogen trace
Supplier:   PEAK SCIENTIFIC
Nitrogen generators, precision nitrogen trace
Supplier:  PEAK SCIENTIFIC
Certificates
Nitrogen generators, precision nitrogen trace
Nitrogen generators, precision nitrogen trace
3 Options Available Below
Product Details & Documents
These precision nitrogen trace generators produce "zero nitrogen" on demand from a compressed air source.
  • Suitable for carrier gas, make up gas and sample preparation
  • Regenerative CMS columns remove oxygen and moisture
  • Catalyst chamber to remove hydrocarbons (as methane) to <0,05 ppm with no expensive catalyst chamber replacements
Maximum pressure: 5,5 bar
Min/ max operating temperature: 5 to 35 °C
Gas outlet: 1×¹/₄" BSPP
Purity: >99,9995%
Weight: 21 kg (26 kg for the 1,00 l/min flow rate model)
Product Family Options
Product Information
  • Type
    Flow rate
    W×D×H
    Availability
    Price
  • Precision Nitrogen Trace
    0,25 l/min
    380×540×256 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 285,00 €
    7 285,00 €
    1 * 1 ST
       7 285,00 €
  • Precision Nitrogen Trace
    0,60 l/min
    380×540×256 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 680,00 €
    8 680,00 €
    1 * 1 ST
       8 680,00 €
  • Precision Nitrogen Trace
    1,00 l/min
    380×540×256 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10 550,00 €
    10 550,00 €
    1 * 1 ST
       10 550,00 €