T7 RNA polymerase
Catalog # EP0111
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Application:
- Description:T7 RNA Polymerase (20 U/µl)
- Environmentally Preferable:
Specifications
About this item
T7 RNA polymerase is ideal for synthesis of unlabeled and labeled RNA. It catalyses the 5'→3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter. It is also used in the study of RNA secondary structure and RNA-protein interactions, RNA splicing.