Order Entry
Austria
ContactUsLinkComponent
Human Recombinant Cathepsin F (from Insect cells)
Human Recombinant Cathepsin F (from Insect cells)
Catalog # ENZOBMLSE5680005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human Recombinant Cathepsin F (from Insect cells)
Catalog # ENZOBMLSE5680005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Enzyme type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Seq:
    02
  • Storage conditions:
    –80 °C
  • Form:
    Liquid
  • Enzyme name:
    Cathepsin F
  • Enzyme activity:
    ≥1 U/mg
  • Purity:
    ≥80%
  • Molecular weight:
    ~39 kDa
  • pH:
    5,0

Specifications

About this item

Recombinant glycosylated procathepsin F cloned from human cDNA (NM_003793), expressed in insect cells, and purified as the active form of the enzyme. Cathepsin F, a member of the papain family of lysosomal cysteine proteases, acts upon proteins such as MHC Class II-associated invariant chain and ApoB-100.

  • Produced in insect cells (secreted) as untagged proenzyme; activated during purification
  • Highly active
  • Purified by multi-step chromatography (SDS-PAGE)
  • Useful tool to study enzyme kinetics, cleave target substrates, and screen for inhibitors

It is a lysosomal protease expressed in numerous tissues, notably brain and macrophages. It functions in the immune system and is implicated in disease states such as atherosclerosis, cancer, and angiogenesis.

It has a liquid formulation in 20 mM NaOAc at pH 5,0, containing 2,5 mM EDTA, 250 mM sodium chloride and 20 mM L-cysteine.

One unit hydrolyses 1 µmol Z-Phe-Arg-AMC substrate (OMNICATHEPSIN® Fluorogenic substrate) per min. at 37 °C.