Order Entry
Austria
ContactUsLinkComponent
 
Human recombinant SIRT5 (from E. coli), His-tag
Catalog # ENZOBMLSE5550050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant SIRT5 (from E. coli), His-tag
Catalog # ENZOBMLSE5550050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 KU
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Storage conditions:
    –80 °C
  • Protein synonyms:
    SIR2L5|NAD-dependent deacetylase Sirtuin-5|Sirtuin 5|Sir2-like protein 5
  • Protein/peptide name:
    SIRT5
  • Purity:
    >95%
  • Molecular weight:
    ~32,3 kDa
  • Formulation:
    Liquid
  • pH:
    7,5

Specifications

About this item

SIRT5, along with SIRTs 3 and 4, is one of three human mitochondrial sirtuins (homologs of yeast Sir2). SIRT5, which is localised to both the mitochondrial matrix and the intermembrane space, is an NAD+-dependent lysine deacetylase (class III HDAC).

  • Produced in E. coli
  • Highly active
  • Purified by multi-step chromatography (SDS-PAGE)
  • Useful for studies of enzyme kinetics, modulator activity, drug discovery

It has a liquid formulation in 25 mM TRIS at pH 7,5, 100 mM sodium chloride, 5 mM DTT, containing 10% (v/v) glycerol.