Order Entry
Austria
ContactUsLinkComponent
Mouse CXCL4 ELISA kit
Mouse CXCL4 ELISA kit
Catalog # BIRBORB106613-96
Supplier:  Biorbyt
CAS Number:  
undefined
Mouse CXCL4 ELISA kit
Catalog # BIRBORB106613-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse CXCL4 ELISA Kit
  • Detection range:
    15,6 - 1000 pg/ml
  • Sensitivity:
    <1 pg/ml

Specifications

About this item

Mouse CXCL4 ELISA Kit for the quantitative determination of mouse chemokine (C-X-C motif) ligand 4 (CXCL4).

Platelet factor 4 (PF4) is a small cytokine belonging to the CXC chemokine family that is also known as chemokine (C-X-C motif) ligand 4 (CXCL4). The PF4 gene was localised on map to 4q12-q13. Chemokines play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis.

PF4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralisation of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair.