Order Entry
Austria
ContactUsLinkComponent
Rat sL-Selectin ELISA kit
Rat sL-Selectin ELISA kit
Catalog # BIRBORB106606-96
Supplier:  Biorbyt
CAS Number:  
undefined
Rat sL-Selectin ELISA kit
Catalog # BIRBORB106606-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Rat
  • Target protein:
    sL-selectin
  • Description:
    Rat sL-Selectin ELISA Kit
  • Detection range:
    625 - 40000 pg/ml
  • Sensitivity:
    <20 pg/ml

Specifications

About this item

Rat sL-Selectin ELISA Kit for the quantitative measurement of rat sL-selectin.

L-selectin, also known as CD62L, is a cell adhesion molecule found on leukocytes. It belongs to the selectin family of proteins, which recognise sialylated carbohydrate groups. It is cleaved by ADAM17.SELL. L-selectin is a cell surface component that is a member of a family of adhesion/homing receptors which play important roles in leukocyte-endothelial cell interactions. The molecule is composed of multiple domains – one homologous to lectins, one to epidermal growth factor, and two to the consensus repeat units found in C3/C4 binding proteins. L-selectin acts as a "homing receptor" for leukocytes to enter secondary lymphoid tissues via high endothelial venules. Ligands present on endothelial cells will bind to leukocyte expressing L-selectin, slowing leukocyte trafficking through the blood, and facilitating entry into a secondary lymphoid organ at that point.