Orders LinkContactUsLinkComponent
Fused quartz microscope slides
Fused quartz microscope slides
Fused quartz microscope slides
Certificates
Fused quartz microscope slides
6 Options Available Below
Product Details & Documents
These fused quartz slides are made of fusing naturally occurring crystalline silica.
  • High chemical purity, high corrosion resistance, high melting point
  • Low coefficient of thermal expansion with excellent electrical insulation
  • Optical transmission from ultra-violet to infrared
Product Family Options
Product Information
  • Material
    L×W
    Availability
    Price
  • 42295.KG
    Microscope slides
    Microscope slides
    Fused quartz
    26×26 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,20 €
    60,20 €
    1 * 1 ST
       60,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    42295.KG
  • Microscope slides
    Fused quartz
    26×51 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,90 €
    68,90 €
    1 * 1 ST
       68,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    42296.KG
  • Microscope slides
    Fused quartz
    76×26 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 78,00 €
    78,00 €
    1 * 1 ST
       78,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    42297.KG
  • 42295.KJ
    Microscope slides
    Microscope slides
    Fused quartz
    26×26 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 209,00 €
    209,00 €
    1 * 5 ST
       209,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    42295.KJ
  • Microscope slides
    Fused quartz
    26×51 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 276,00 €
    276,00 €
    1 * 5 ST
       276,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    42296.KJ
  • Microscope slides
    Fused quartz
    76×26 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 312,00 €
    312,00 €
    1 * 5 ST
       312,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    42297.KJ