Order Entry
Austria
ContactUsLinkComponent
Buffer, PBS tablets
Buffer, PBS tablets
Catalog # A9177.0100
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Buffer, PBS tablets
Catalog # A9177.0100
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    100
  • Description:
    PBS tablets, pH 7,4

Specifications

About this item

PBS Tablets for preparation of 200 ml phosphate buffered saline (PBS). Composition of 140 mM sodium chloride, 2,7 mM potassium chloride and 10 mM phosphate.

  • Ready to use in minutes
  • Autoclavable
  • Stable for approx. 12 months if stored at 2 to 8 °C

Deposit one tablet in flask or beaker placed on a magnetic stirrer. Add deionised water to 50% of the final volume at room temperature. Stir for a few minutes. Adjust the water to the final volume, stir until full dissolution. Autoclave or filter sterilise through a 0,22 µm filter into a sterile flask or bottle.