Order Entry
Austria
ContactUsLinkComponent
Oxidation stability reactor, OXITEST
Oxidation stability reactor, OXITEST
Catalog # 705-0862
Supplier:  VELP SCIENTIFIC
undefined
Oxidation stability reactor, OXITEST
Catalog # 705-0862
Supplier:  VELP SCIENTIFIC

Specifications

  • Sample size:
    Max. 100 ml
  • Temperature range:
    Room temp. to 110 °C
  • Description:
    OXITEST oxidation stability reactor
  • Electrical:
    230 V / 50- 60 Hz
    900 W
  • W×D×H:
    365 x 190 x 485 mm
  • Weight:
    16,5 kg
  • Pk:
    1

Specifications

About this item

Innovative accelerated oxidation test for the determination of the oxidation stability of fats on the whole sample according to AOCS Cd-12c-16.

  • Titanium sample holders offer excellent chemical resistance and temperature homogeneity
  • Two separate titanium chambers in order to test the same sample in duplicate or different samples at the same time
  • PC control for representative results; programmable for unattended operation
  • Suitable for oxidation stability in foods, oils and fats
  • Analysis on the whole sample, without preliminary fat separation
  • Accelerated test, obtained in a relatively short period of time
  • Oxidation stability without the need of expensive and environmental hazardous reagents

OXITEST is a multi-application accelerated oxidative stability testing device. Multiple application - prediction of oxidative stability during shelf-life studies. Tests the product at defined time intervals; builds an experimental curve. Evaluates the adequacy of storage conditions and the best packaging solution. Compares the oxidative stability of different formulas for food preparations. Evaluates the oxidative stability of vegetable oils of different botanical origin also the effectiveness of antioxidants. Informs on product oxidation when the oxidation flex is not visible. Especially useful for products with a low fat content (4 to 5%). Product oxidation can be achieved by combining the OXITEST with GC. Can offer an oxidative stability test of incoming raw materials and outgoing finished products.

Simple and safe operationally intuitive OXISoft™ software; easy to learn and use. Over-pressure safety valve. Alarms for damaged probe and 'out of range' temperature. Choice of language for the management software.

Versatile data handling – data recordable in a database. Test comparisons available with data overlays. Data exporting via USB to pen drive or PC. Output in .xls, .txt and .csv format to match reporting systems.

Robust design – indestructible titanium sample holders and chambers. Hermetically sealed closure of the chambers.

Delivery: OXITEST is supplied with OXISoft™ software, USB cable, 6 space holders and 4 spacers.