Order Entry
Austria
ContactUsLinkComponent
Section lifter, flexible
Section lifter, flexible
Catalog # BOCH3537
Supplier:  BOCHEM
undefined
Section lifter, flexible
Catalog # BOCH3537
Supplier:  BOCHEM

Specifications

  • Length:
    150 mm
  • Blade width:
    20 mm
  • Pk:
    1
  • Blade length:
    50 mm

Specifications

About this item

Section lifter with flexible stainless steel blade and wooden handle.