ContactUsLinkComponent
Qualitative filter papers, wet strengthened grades, grade 1574 and 1574 ½, Whatman™
Qualitative filter papers, wet strengthened grades, grade 1574 and 1574 ½, Whatman™
Certificates
undefined
Qualitative filter papers, wet strengthened grades, grade 1574 and 1574 ½, Whatman™
Specifications
  • Catalog No:
  • 512-0646
  • WHAT10314828
  • 515-7104
  • 515-7103
  • Grade:
  • 1574
  • 1574
  • 1574 ½
  • 1574 ½
  • Weight:
  • 90 g/m²
  • 90 g/m²
  • 90 g/m²
  • 90 g/m²
  • Thickness:
  • 160 μm
  • 160 μm
  • 160 μm
  • 160 μm
  • Particle retention:
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • Filtration speed:
  • 85 s (Herzberg)
  • 85 s (Herzberg)
  • 85 s (Herzberg)
  • 85 s (Herzberg)