Mouse recombinant IFN beta (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Avoid multiple freeze-thaw cycles.
- Protein synonyms:beta-interferon
- Protein/peptide name:IFN beta
- Purity:Greater than 95% as determined by Analysis by SDS-PAGE.
- Sequence:MGSSHHHHHH SSGLVPRGSH MINYKQLQLQ ERTNIRKCQE LLEQLNGKIN LTYRADFKIP MEMTEKMQKS YTAFAIQEML QNVFLVFRNN FSSTGWNETI VVRLLDELHQ QTVFLKTVLE EKQEERLTWE MSSTALHLKS YYWRVQRYLK LMKYNSYAWM VVRAEIFRNF LIIRRLTRNF QN.
- Formulation:The IFN-Beta contains 20 mM Tris-HCl buffer pH-8, 0.1M NaCl and 30% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together