Human recombinant CTLA4 (from Insect cells (Sf9))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Storage conditions:Lyophilized Cytotoxic T-Lymphocyte Associated Antigen Recombinant although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution CTLA-4 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:ALPS5
- Protein/peptide name:CTLA4
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Cytotoxic T-Lymphocyte Associated Antigen-4 Human in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the amino acids was determined and was found to be: Lys-Ala-Met-His-Val.
- Formulation:CTLA-4 was lyophilized with no additives.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together