Human Eukaryotic Translation Initiation Factor 4E (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Avoid multiple freeze-thaw cycles.
- Protein synonyms:EIF4E1
- Protein/peptide name:Eukaryotic Translation Initiation Factor 4E
- Purity:Greater than 85.0% as determined by SDS-PAGE.
- Sequence:MGSSHHHHHH SSGLVPRGSH MATVEPETTP TPNPPTTEEE KTESNQEVAN PEHYIKHPLQ NRWALWFFKN DKSKTWQANL RLISKFDTVEDFWALYNHIQ LSSNLMPGCD YSLFKDGIEP MWEDEKNKRG GRWLITLNKQ QRRSDLDRFW LETLLCLIGE SFDDYSDDVC GAVVNVRAKG DKIAIWTTEC ENREAVTHIG RVYKERLGLP PKIVIGYQSH ADTATKSGST TKNRFVV.
- Formulation:The EIF4E 0.5mg/ml protein solution contains 20 mM Tris-HCl, pH-8, and 10% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together