Human Troponin I-C (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Troponin IC although stable at 10 °C for days, should be stored desiccated below −18 °C. Please prevent freeze-thaw cycles.
- Protein synonyms:Troponin cardiac muscle
- Protein/peptide name:Troponin I-C
- Purity:Greater than 90.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Formulation:Each mg/ml of protein (0.5mg/ml) contains 50% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together