Mouse VEGF-Sf9 (from Baculovirus (Sf9 Insect cells))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Baculovirus (Sf9 Insect cells)
- Species:Mouse
- Storage conditions:Lyophilized Vascular Endothelial Growth Factor Sf9 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution VEGF-Sf9 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Biological activity:The ED50 range, determined by the dose-dependent proliferation of human umbilical vein endothelial cells (HUVEC) (measured by 3H-thymidine uptake) is 1-2 ng/ml, corresponding to specific activity of 1,000,000 Units/mg.
- Protein synonyms:Vascular endothelial growth factor A
- Protein/peptide name:VEGF-Sf9
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Vascular Endothelial Growth Factor-Sf9 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:The protein was lyophilized from concentrated (1mg/ml) solution with no additives.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together