Order Entry
Austria
ContactUsLinkComponent
Encephalitis C-end
Encephalitis C-end
Supplier:  Biorbyt
Certificates
Encephalitis C-end
Supplier:  Biorbyt

Specifications

  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    Encephalitis C-end
  • Purity:
    Encephalitis protein is >95% pure as determined by 10% PAGE (coomassie staining). Encephalitis protein was purified by proprietary chromatographic technique.
  • Formulation:
    20 mM MES pH 6.5, 8M urea, 200 mM NaCL and 0.05% Tween-20.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together